(S)-2-Amino-2-(3-fluoro-5-methoxyphenyl)ethanol 97%

Supplier: Ambeed
Warning

A926565-100MG A926565-5G
76655-508EA 79.57 USD
76655-508 76655-510
(S)-2-Amino-2-(3-fluoro-5-methoxyphenyl)ethanol 97%
(S)-2-Amino-2-(3-fluoro-5-methoxyphenyl)ethanol

Precaución: for Research Use Only

Formula: C₉H₁₂FNO₂
MW: 185.1955 g/mol
MDL Number: MFCD09415636
CAS Number: 1212981-84-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR