1,3-Bis(2-propynyloxy)benzene 97% (by GC)

Supplier: Ambeed
Warning

Synonyms: 1,3-Bis(propargyloxy)benzene

A924905-100MG A924905-1G A924905-250MG
76655-502EA 29.71 USD
76655-502 76655-504 76655-506
1,3-Bis(2-propynyloxy)benzene 97% (by GC)
1,3-Bis(2-propynyloxy)benzene

Precaución: for Research Use Only

Formula: C₁₂H₁₀O₂
MW: 186.21 g/mol
MDL Number: MFCD00224312
CAS Number: 26627-36-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR