rel-(1R,2R,4S)-7-oxabicyclo[2.2.1]heptane-2-carboxylic acid 98+%

Supplier: Ambeed
Warning

A924350-100MG A924350-1G A924350-250MG
76655-490EA 120.95 USD
76655-490 76655-492 76655-494
rel-(1R,2R,4S)-7-oxabicyclo[2.2.1]heptane-2-carboxylic acid 98+%
rel-(1R,2R,4S)-7-oxabicyclo[2.2.1]heptane-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₇H₁₀O₃
MW: 142.1525 g/mol
CAS Number: 38263-55-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR