3-Chlorobicyclo[1.1.1]pentan-1-amine hydrochloride 95%

Supplier: Ambeed
Warning

A924258-1G A924258-250MG A924258-100MG A924258-50MG
76655-486EA 2126.05 USD
76655-486 76655-488 77618-572 77618-574
3-Chlorobicyclo[1.1.1]pentan-1-amine hydrochloride 95%
3-Chlorobicyclo[1.1.1]pentan-1-amine hydrochloride

Precaución: for Research Use Only

Formula: C₅H₉Cl₂N
MW: 154.04 g/mol
MDL Number: MFCD30803728
CAS Number: 1886967-25-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR