BAM15 ≥99%

Supplier: Ambeed
Danger

Synonyms: BAM 15, N5,N6-Bis(2-fluorophenyl)-[1,2,5]oxadiazolo[3,4-b]pyrazine-5,6-diamine

A924230-10MG A924230-1MG A924230-25MG A924230-5MG
76655-478EA 207.94 USD
76655-478 76655-480 76655-482 76655-484
BAM15 ≥99%
BAM15

Precaución: for Research Use Only

Formula: C₁₆H₁₀F₂N₆O
MW: 340.29 g/mol
MDL Number: MFCD00373912
CAS Number: 210302-17-3
UN: 3077
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR