Polyethylene glycol monomethyl ether, M.W. ∼400
Supplier: Ambeed
|
Warning
|
Synonyms:
Methoxypolyethylene glycols, Methoxy poly(ethylene glycol), PEG MME, Poly(ethylene glycol) methyl ether, mPEG, Methoxypolyethylene glycols-average M.W. 1000, m-PEG-OH
A924038-1000G
A924038-100G
A924038-25G
A924038-500G
76655-470EA
41
USD
76655-470
76655-472
76655-474
76655-476
Polyethylene glycol monomethyl ether, M.W. ∼400
Polyethylene glycol monomethyl ether
Precaución: for Research Use Only
Formula:
CH₃(OCH₂CH₂)nOH |
MDL Number:
MFCD00084416 CAS Number: 9004-74-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...