(S)-2-Morpholin-4-yl-propionic acid hydrochloride 97%

Supplier: Ambeed
Warning

A923890-100MG A923890-1G A923890-250MG
76655-464EA 67.9 USD
76655-464 76655-466 76655-468
(S)-2-Morpholin-4-yl-propionic acid hydrochloride 97%
(S)-2-Morpholin-4-yl-propionic acid hydrochloride

Precaución: for Research Use Only

Formula: C₇H₁₄ClNO₃
MW: 195.6400 g/mol
MDL Number: MFCD22689280
CAS Number: 237427-96-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR