2,5-Dimethoxy-4-propylbenzaldehyde 95%

Supplier: Ambeed
Warning

A923143-100MG A923143-1G A923143-250MG A923143-5G
76655-446EA 44 USD
76655-446 76655-448 76655-450 76655-452
2,5-Dimethoxy-4-propylbenzaldehyde 95%
2,5-Dimethoxy-4-propylbenzaldehyde

Precaución: for Research Use Only

Formula: C₁₂H₁₆O₃
MDL Number: MFCD19302282
CAS Number: 53581-81-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR