Naphthalene-1,2-diamine dihydrochloride 95%

Supplier: Ambeed
Warning

A923074-1G A923074-250MG
76655-442EA 57 USD
76655-442 76655-444
Naphthalene-1,2-diamine dihydrochloride 95%
Naphthalene-1,2-diamine dihydrochloride

Precaución: for Research Use Only

Formula: C₁₀H₁₂Cl₂N₂
MW: 231.1217 g/mol
MDL Number: MFCD01596273
CAS Number: 78196-74-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR