Benzyl (2-(2-(2-bromoethoxy)ethoxy)ethyl)carbamate 98%

Supplier: Ambeed
Warning

Synonyms: Cbz-PEG2-bromide

A1350032-100MG A1350032-250MG A1350032-1G A1350032-5G
76658-172EA 44 USD
76658-172 76658-174 77549-622 77621-632
Benzyl (2-(2-(2-bromoethoxy)ethoxy)ethyl)carbamate 98%
Benzyl (2-(2-(2-bromoethoxy)ethoxy)ethyl)carbamate

Precaución: for Research Use Only

Formula: C₁₄H₂₀BrNO₄
MW: 346.2169 g/mol
MDL Number: MFCD31916729
CAS Number: 2100283-00-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR