(S)-2-(But-3-en-1-yl)oxirane 98%

Supplier: Ambeed
Danger

A921350-25G A921350-5G
76655-404EA 321.46 USD
76655-404 76655-406
(S)-2-(But-3-en-1-yl)oxirane 98%
(S)-2-(But-3-en-1-yl)oxirane

Precaución: for Research Use Only

Formula: C₆H₁₀O
MW: 98.14 g/mol
MDL Number: MFCD17926427
CAS Number: 137688-21-2
UN: 1992
ADR: 3,6.1,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR