5-Amino-1H-pyrazol-3-ol 95%

Supplier: Ambeed
Warning

A920251-1G A920251-25G A920251-5G
76655-380EA 7.43 USD
76655-380 76655-382 76655-384
5-Amino-1H-pyrazol-3-ol 95%
5-Amino-1H-pyrazol-3-ol

Precaución: for Research Use Only

Formula: C₃H₅N₃O
MW: 99.0913 g/mol
MDL Number: MFCD00233932
CAS Number: 145092-03-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR