4-Benzylpiperidine-1,4-dicarboxylic acid mono-tert-butyl ester 97%

Supplier: Ambeed
Warning

Synonyms: 4-Benzyl-1-(tert-butoxycarbonyl)piperidine-4-carboxylic acid, 1-Boc-4-benzyl-4-piperidinecarboxylic acid

A919730-100MG A919730-250MG A919730-1G
76655-378EA 45 USD
76655-378 77623-146 77784-355
4-Benzylpiperidine-1,4-dicarboxylic acid mono-tert-butyl ester 97%
4-Benzylpiperidine-1,4-dicarboxylic acid mono-tert-butyl ester

Precaución: for Research Use Only

Formula: C₁₈H₂₅NO₄
MDL Number: MFCD02179131
CAS Number: 167263-11-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR