(R)-5,6,7,8-tetrahydroquinolin-8-amine hydrochloride 95%
Supplier: Ambeed
|
Warning
|
A917901-100MG
A917901-1G
A917901-250MG
76655-346EA
36
USD
76655-346
76655-348
76655-350
(R)-5,6,7,8-tetrahydroquinolin-8-amine hydrochloride 95%
(R)-5,6,7,8-tetrahydroquinolin-8-amine hydrochloride
Precaución: for Research Use Only
Formula:
C₉H₁₂N₂·ClH |
MDL Number:
MFCD31654063 CAS Number: 1431726-92-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...