2-(4-(Benzyloxy)-3,5-dimethylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 97%

Supplier: Ambeed
Warning

A917709-1G A917709-250MG A917709-5G
76655-340EA 83.82 USD
76655-340 76655-342 76655-344
2-(4-(Benzyloxy)-3,5-dimethylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 97%
2-(4-(Benzyloxy)-3,5-dimethylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

Precaución: for Research Use Only

Formula: C₂₁H₂₇BO₃
MW: 338.2483 g/mol
MDL Number: MFCD23699512
CAS Number: 2121513-92-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR