4-(Oxiran-2-ylmethoxy)benzeneboronic acid, pinacol ester 98%

Supplier: Ambeed
Warning

A917035-100MG A917035-1G A917035-250MG
76655-332EA 37 USD
76655-332 76655-334 76655-336
4-(Oxiran-2-ylmethoxy)benzeneboronic acid, pinacol ester 98%
4-(Oxiran-2-ylmethoxy)benzeneboronic acid, pinacol ester

Precaución: for Research Use Only

Formula: C₁₅H₂₁BO₄
MDL Number: MFCD09027286
CAS Number: 664991-83-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR