(3S,5S)-3,5-Dimethylpiperazin-2-one 97%

Supplier: Ambeed
Warning

A916739-100MG A916739-1G A916739-250MG A916739-50MG
76655-324EA 102 USD
76655-324 76655-326 76655-328 76655-330
(3S,5S)-3,5-Dimethylpiperazin-2-one 97%
(3S,5S)-3,5-Dimethylpiperazin-2-one

Precaución: for Research Use Only

Formula: C₆H₁₂N₂O
MDL Number: MFCD18250093
CAS Number: 1152112-99-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR