1-Amino-3,6,9,12,15,18,21,24-octaoxaheptacosan-27-oic acid 97%

Supplier: Ambeed
Warning

Synonyms: NH2-PEG8-acid, α-Amine-ω-propionic acid octaethylene glycol, H2N-PEG8-CH2CH2COOH

A916666-250MG A916666-1G A916666-5G
76655-322EA 69 USD
76655-322 77603-294 77784-349
1-Amino-3,6,9,12,15,18,21,24-octaoxaheptacosan-27-oic acid 97%
1-Amino-3,6,9,12,15,18,21,24-octaoxaheptacosan-27-oic acid

Precaución: for Research Use Only

Formula: C₁₉H₃₉NO₁₀
MDL Number: MFCD11041146
CAS Number: 756526-04-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR