Isopropylamine hydrobromide 98% (by total nitrogen and titration analysis)

Supplier: Ambeed
Warning

Synonyms: Isopropylammonium bromide

A916647-1G A916647-25G A916647-5G
76655-316EA 16.98 USD
76655-316 76655-318 76655-320
Isopropylamine hydrobromide 98% (by total nitrogen and titration analysis)
Isopropylamine hydrobromide

Precaución: for Research Use Only

Formula: C₃H₉N·HBr
MW: 140.02 g/mol
MDL Number: MFCD22419143
CAS Number: 29552-58-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR