4-(4-Trifluoromethylbenzoyl)piperidine hydrochloride 95%

Supplier: Ambeed
Warning

A916431-100MG A916431-250MG
76655-312EA 26 USD
76655-312 76655-314
4-(4-Trifluoromethylbenzoyl)piperidine hydrochloride 95%
4-(4-Trifluoromethylbenzoyl)piperidine hydrochloride

Precaución: for Research Use Only

Formula: C₁₃H₁₅ClF₃NO
MDL Number: MFCD04039702
CAS Number: 25519-83-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR