tert-Butyl N-[2-(4-nitrophenyl)ethyl]carbamate 98%

Supplier: Ambeed
Warning

A916092-10G A916092-1G A916092-250MG A916092-5G
76655-302EA 183.54 USD
76655-302 76655-304 76655-306 76655-308
tert-Butyl N-[2-(4-nitrophenyl)ethyl]carbamate 98%
tert-Butyl N-[2-(4-nitrophenyl)ethyl]carbamate

Precaución: for Research Use Only

Formula: C₁₃H₁₈N₂O₄
MW: 266.2930 g/mol
MDL Number: MFCD16620906
CAS Number: 144226-16-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR