Sr 8278 98%

Supplier: Ambeed
Warning

Synonyms: Ethyl 2-(5-(methylthio)thiophene-2-carbonyl)-1,2,3,4-tetrahydroisoquinoline-3-carboxylate, SR8278

A915123-10MG A915123-1MG A915123-25MG A915123-50MG A915123-5MG
76655-280EA 55 USD
76655-280 76655-282 76655-284 76655-286 76655-288
Sr 8278 98%
Sr 8278

Precaución: for Research Use Only

Formula: C₁₈H₁₉NO₃S₂
MDL Number: MFCD18782736
CAS Number: 1254944-66-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR