4-(Bromomethyl)benzo[d]thiazole 95%

Supplier: Ambeed
Danger

A913545-250MG A913545-100MG
76655-268EA 260.99 USD
76655-268 77749-727
4-(Bromomethyl)benzo[d]thiazole 95%
4-(Bromomethyl)benzo[d]thiazole

Precaución: for Research Use Only

Formula: C₈H₆BrNS
MW: 228.1089 g/mol
MDL Number: MFCD22571526
CAS Number: 936827-73-5
UN: 3261
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR