4-(3-(4-Hydroxyphenyl)-4,4-dimethyl-5-oxo-2-thioxoimidazolidin-1-yl)-2-(trifluoromethyl)benzonitrile 98%
Supplier: Ambeed
|
Warning
|
A913149-100MG
A913149-1G
A913149-250MG
A913149-5G
76655-252EA
33.95
USD
76655-252
76655-254
76655-256
76655-258
4-(3-(4-Hydroxyphenyl)-4,4-dimethyl-5-oxo-2-thioxoimidazolidin-1-yl)-2-(trifluoromethyl)benzonitrile 98%
4-(3-(4-Hydroxyphenyl)-4,4-dimethyl-5-oxo-2-thioxoimidazolidin-1-yl)-2-(trifluoromethyl)benzonitrile
Precaución: for Research Use Only
Formula:
C₁₉H₁₄F₃N₃O₂S MW: 405.3936 g/mol |
MDL Number:
MFCD28978634 CAS Number: 915086-32-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...