1,3,5-Tris[4-(diphenylamino)phenyl]benzene 95% (by total nitrogen basis)

Supplier: Ambeed
Warning

A910621-100MG A910621-1G A910621-250MG
76655-234EA 27 USD
76655-234 76655-236 76655-238
1,3,5-Tris[4-(diphenylamino)phenyl]benzene 95% (by total nitrogen basis)
1,3,5-Tris[4-(diphenylamino)phenyl]benzene

Precaución: for Research Use Only

Formula: C₆₀H₄₅N₃
MDL Number: MFCD30721946
CAS Number: 147951-36-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR