tert-Butyl 11-bromoundecanoate 97%

Supplier: Ambeed
Warning

A910456-100MG A910456-1G A910456-250MG A910456-5G A910456-25G
76655-226EA 48.81 USD
76655-226 76655-228 76655-230 76655-232 77618-542
tert-Butyl 11-bromoundecanoate 97%
tert-Butyl 11-bromoundecanoate

Precaución: for Research Use Only

Formula: C₁₅H₂₉BrO₂
MW: 321.2936 g/mol
MDL Number: MFCD31727846
CAS Number: 85216-74-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR