3-Methoxy-N-(4H-1,2,4-triazol-4-yl)benzamide 90%

Supplier: Ambeed
Warning

A909983-100MG A909983-1G A909983-250MG
76655-220EA 24 USD
76655-220 76655-222 76655-224
3-Methoxy-N-(4H-1,2,4-triazol-4-yl)benzamide 90%
3-Methoxy-N-(4H-1,2,4-triazol-4-yl)benzamide

Precaución: for Research Use Only

Formula: C₁₀H₁₀N₄O₂
MDL Number: MFCD00750819
CAS Number: 331434-10-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR