Ethyl 2-amino-3,4,5-trifluorobenzoate 97%

Supplier: Ambeed
Danger

A907888-100MG A907888-1G A907888-250MG
76655-212EA 13 USD
76655-212 76655-214 76655-216
Ethyl 2-amino-3,4,5-trifluorobenzoate 97%
Ethyl 2-amino-3,4,5-trifluorobenzoate

Precaución: for Research Use Only

Formula: C₉H₈F₃NO₂
MDL Number: MFCD12159863
CAS Number: 1184500-46-6
UN: 2810
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR