2-(2-Chloro-4-methoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 97%

Supplier: Ambeed
Warning

Synonyms: 2-Chloro-4-methoxyphenylboronic acid pinacol ester

A907452-1G A907452-250MG A907452-5G
76655-206EA 76.39 USD
76655-206 76655-208 76655-210
2-(2-Chloro-4-methoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 97%
2-(2-Chloro-4-methoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

Precaución: for Research Use Only

Formula: C₁₃H₁₈BClO₃
MW: 268.54 g/mol
MDL Number: MFCD22414622
CAS Number: 1416722-82-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR