2,6-Difluoro-4-(trifluoromethyl)benzoic acid 97%

Supplier: Ambeed
Warning

Synonyms: α,α,α,2,6-Pentafluoro-p-toluic acid, 4-Carboxy-3,5-difluorobenzotrifluoride

A906647-100MG A906647-1G A906647-250MG
76655-190EA 67.9 USD
76655-190 76655-192 76655-194
2,6-Difluoro-4-(trifluoromethyl)benzoic acid 97%
2,6-Difluoro-4-(trifluoromethyl)benzoic acid

Precaución: for Research Use Only

Formula: C₈H₃F₅O₂
MW: 226.1 g/mol
MDL Number: MFCD27975469
CAS Number: 1309597-24-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR