1-Hexylpyridinium bromide 99%

Supplier: Ambeed
Warning

A906271-100G A906271-25G A906271-5G
76655-182EA 59 USD
76655-182 76655-184 77618-534
1-Hexylpyridinium bromide 99%
1-Hexylpyridinium bromide

Precaución: for Research Use Only

Formula: C₁₁H₁₈BrN
MDL Number: MFCD03427613
CAS Number: 74440-81-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR