2-(3,5-Dimethoxybenzoyl)-N-(naphthalen-1-yl)hydrazinecarboxamide 98%
Supplier: Ambeed
|
Warning
|
A905779-10MG
A905779-1MG
A905779-25MG
A905779-5MG
76655-164EA
35
USD
76655-164
76655-166
76655-168
76655-170
2-(3,5-Dimethoxybenzoyl)-N-(naphthalen-1-yl)hydrazinecarboxamide 98%
2-(3,5-Dimethoxybenzoyl)-N-(naphthalen-1-yl)hydrazinecarboxamide
Precaución: for Research Use Only
Formula:
C₂₀H₁₉N₃O₄ |
MDL Number:
MFCD02372953 CAS Number: 444932-31-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...