4-(tetramethyl-1,3,2-dioxaborolan-2-yl)naphthalene-1-carbonitrile 97%

Supplier: Ambeed
Warning

A905722-100MG A905722-1G A905722-250MG
76655-158EA 23 USD
76655-158 76655-160 76655-162
4-(tetramethyl-1,3,2-dioxaborolan-2-yl)naphthalene-1-carbonitrile 97%
4-(tetramethyl-1,3,2-dioxaborolan-2-yl)naphthalene-1-carbonitrile

Precaución: for Research Use Only

Formula: C₁₇H₁₈BNO₂
MDL Number: MFCD28167500
CAS Number: 1352794-90-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR