Phenyl N-hydroxycarbamate 97%

Supplier: Ambeed
Warning

A905369-250MG A905369-1G A905369-5G
76655-156EA 28 USD
76655-156 77573-822 77623-134
Phenyl N-hydroxycarbamate 97%
Phenyl N-hydroxycarbamate

Precaución: for Research Use Only

Formula: C₇H₇NO₃
MDL Number: MFCD18968936
CAS Number: 38064-07-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR