(8-Methylquinolin-6-yl)boronic acid 96%

Supplier: Ambeed
Warning

A903307-100MG A903307-1G A903307-250MG
76655-122EA 39 USD
76655-122 76655-124 76655-126
(8-Methylquinolin-6-yl)boronic acid 96%
(8-Methylquinolin-6-yl)boronic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₀BNO₂
MDL Number: MFCD27935558
CAS Number: 2260683-75-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR