3-Bromo-2-methylbenzenemethanamine 95%

Supplier: Ambeed
Warning

A900457-100MG A900457-250MG
76655-110EA 53.05 USD
76655-110 76655-112
3-Bromo-2-methylbenzenemethanamine 95%
3-Bromo-2-methylbenzenemethanamine

Precaución: for Research Use Only

Formula: C₈H₁₀BrN
MW: 200.0757 g/mol
MDL Number: MFCD18392010
CAS Number: 943722-02-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR