Lithium bromide hydrate 99%
Supplier: Ambeed
|
Warning
|
Synonyms:
Lithium bromide xhydrate
A900242-100G
A900242-25G
A900242-500G
A900242-5G
76655-102EA
26
USD
76655-102
76655-104
76655-106
76655-108
Lithium bromide hydrate 99%
Lithium bromide hydrate
Precaución: for Research Use Only
Formula:
BrLi·xH₂O MW: 86.85 g/mol (anhy) Boiling Pt: 1265 Melting Pt: 549 |
MDL Number:
MFCD00149763 CAS Number: 85017-82-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...