1-(2-Propynyloxy)naphthalene 95% (by GC)

Supplier: Ambeed
Warning

Synonyms: 1-(Propargyloxy)naphthalene

A899216-250MG A899216-1G A899216-100MG A899216-5G
76655-086EA 36 USD
76655-086 77623-128 77784-332 77784-333
1-(2-Propynyloxy)naphthalene 95% (by GC)
1-(2-Propynyloxy)naphthalene

Precaución: for Research Use Only

Formula: C₁₃H₁₀O
MDL Number: MFCD03787730
CAS Number: 18542-45-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR