5,7-Dichloro-1H-[1,2,3]triazolo[4,5-b]pyridine 95%

Supplier: Ambeed
Warning

A898992-1G A898992-100MG
76655-084EA 546.37 USD
76655-084 77749-706
5,7-Dichloro-1H-[1,2,3]triazolo[4,5-b]pyridine 95%
5,7-Dichloro-1H-[1,2,3]triazolo[4,5-b]pyridine

Precaución: for Research Use Only

Formula: C₅H₂Cl₂N₄
MW: 189.0022 g/mol
CAS Number: 2091505-12-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR