CaCCinh-A01 98%

Supplier: Ambeed
Warning

Synonyms: TMEM16-Blocker I, 6-(1,1-dimethylethyl)-2-[(2-furanylcarbonyl)amino]-4,5,6,7-tetrahydro-benzo[b]thiophene-3-carboxylic acid, 6-(tert-Butyl)-2-(furan-2-carboxamido)-4,5,6,7-tetrahydrobenzo[b]thiophene-3-carboxylic acid

A897954-10MG A897954-1MG A897954-25MG A897954-5MG
76655-076EA 235.52 USD
76655-076 76655-078 76655-080 76655-082
CaCCinh-A01 98%
CaCCinh-A01

Precaución: for Research Use Only

Formula: C₁₈H₂₁NO₄S
MW: 347.43 g/mol
MDL Number: MFCD02919058
CAS Number: 407587-33-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR