(4-Methanesulfonyl-3-methylphenyl)boronic acid 98%

Supplier: Ambeed
Warning

A897029-100MG A897029-1G A897029-250MG
76655-066EA 37 USD
76655-066 76655-068 76655-070
(4-Methanesulfonyl-3-methylphenyl)boronic acid 98%
(4-Methanesulfonyl-3-methylphenyl)boronic acid

Precaución: for Research Use Only

Formula: C₈H₁₁BO₄S
MDL Number: MFCD27935684
CAS Number: 1598437-51-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR