3-(Difluoromethyl)pyridin-2(1H)-one 95%

Supplier: Ambeed
Warning

A897005-100MG A897005-1G A897005-250MG
76655-060EA 198.39 USD
76655-060 76655-062 76655-064
3-(Difluoromethyl)pyridin-2(1H)-one 95%
3-(Difluoromethyl)pyridin-2(1H)-one

Precaución: for Research Use Only

Formula: C₆H₅F₂NO
MW: 145.1068 g/mol
MDL Number: MFCD31699335
CAS Number: 1806766-03-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR