(4-Chloro-2-fluoro-6-methoxyphenyl)boronic acid 97%

Supplier: Ambeed
Warning

A892287-100MG A892287-1G A892287-250MG
76655-024EA 32 USD
76655-024 76655-026 76655-028
(4-Chloro-2-fluoro-6-methoxyphenyl)boronic acid 97%
(4-Chloro-2-fluoro-6-methoxyphenyl)boronic acid

Precaución: for Research Use Only

Formula: C₇H₇BClFO₃
MDL Number: MFCD30329340
CAS Number: 1628684-10-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR