2-Chloro-N-(4-(1,1,1,3,3,3-hexafluoro-2-hydroxypropan-2-yl)phenyl)benzamide 98%

Supplier: Ambeed
Warning

A892129-100MG A892129-10MG A892129-1MG A892129-25MG A892129-50MG A892129-5MG
76655-012EA 402.09 USD
76655-012 76655-014 76655-016 76655-018 76655-020 76655-022
2-Chloro-N-(4-(1,1,1,3,3,3-hexafluoro-2-hydroxypropan-2-yl)phenyl)benzamide 98%
2-Chloro-N-(4-(1,1,1,3,3,3-hexafluoro-2-hydroxypropan-2-yl)phenyl)benzamide

Precaución: for Research Use Only

Formula: C₁₆H₁₀ClF₆NO₂
MW: 397.6995 g/mol
MDL Number: MFCD02693203
CAS Number: 303126-97-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR