1-Methyl-1-propylpiperidinium bromide 98% (by titrimetric analysis)

Supplier: Ambeed
Warning

A891538-100G A891538-25G A891538-5G
76655-000EA 215 USD
76655-000 76655-002 76655-004
1-Methyl-1-propylpiperidinium bromide 98% (by titrimetric analysis)
1-Methyl-1-propylpiperidinium bromide

Precaución: for Research Use Only

Formula: C₉H₂₀BrN
MDL Number: MFCD17014743
CAS Number: 88840-42-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR