1,4-Bis(10-undecenyloxy)benzene 97% (by HPLC)
Supplier: Ambeed
|
Warning
|
A890945-100MG
A890945-1G
76654-994EA
34
USD
76654-994
77552-674
1,4-Bis(10-undecenyloxy)benzene 97% (by HPLC)
1,4-Bis(10-undecenyloxy)benzene
Precaución: for Research Use Only
Formula:
C₂₈H₄₆O₂ |
MDL Number:
MFCD18207714 CAS Number: 138551-10-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...