N-Succinimidyl Methacrylate 98% (by GC)

Supplier: Ambeed
Warning

Synonyms: N-Methacryloyloxysuccinimide, Methacrylic Acid N-Succinimidyl Ester

A890342-1G A890342-5G
76654-984EA 26 USD
76654-984 76654-986
N-Succinimidyl Methacrylate 98% (by GC)
N-Succinimidyl Methacrylate

Precaución: for Research Use Only

Formula: C₈H₉NO₄
MW: 183.16 g/mol
MDL Number: MFCD15072193
CAS Number: 38862-25-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR