4-(Bromomethyl)-3-methylpyridine hydrobromide 95%

Supplier: Ambeed
Danger

A889976-100MG A889976-250MG A889976-1G
76654-980EA 31 USD
76654-980 76654-982 77749-696
4-(Bromomethyl)-3-methylpyridine hydrobromide 95%
4-(Bromomethyl)-3-methylpyridine hydrobromide

Precaución: for Research Use Only

Formula: C₇H₉Br₂N
MDL Number: MFCD30527969
CAS Number: 2201551-69-9
UN: 3261
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR