7-Oxooctanoic acid 96%

Supplier: Ambeed
Warning

A889383-100MG A889383-1G A889383-250MG A889383-5G
76654-972EA 26.53 USD
76654-972 76654-974 76654-976 76654-978
7-Oxooctanoic acid 96%
7-Oxooctanoic acid

Precaución: for Research Use Only

Formula: C₈H₁₄O₃
MW: 158.2 g/mol
MDL Number: MFCD00075271
CAS Number: 14112-98-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR