5,5'-Dibromo-3,3'-dihexyl-2,2'-bithiophene 97%

Supplier: Ambeed
Warning

A888610-1G A888610-250MG A888610-5G
76654-966EA 176.11 USD
76654-966 76654-968 76654-970
5,5'-Dibromo-3,3'-dihexyl-2,2'-bithiophene 97%
5,5'-Dibromo-3,3'-dihexyl-2,2'-bithiophene

Precaución: for Research Use Only

Formula: C₂₀H₂₈Br₂S₂
MW: 492.37 g/mol
MDL Number: MFCD23380207
CAS Number: 170702-05-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR